Would you like to log in again?
Detergents, alkaline, neodisher® MA
Supplier: Miele
neodisher® MA is a mildly alkaline formulation (powder) with de-foaming properties. It removes blood, protein, nutrient mediums, residues of baby food and chemicals from surfaces of all types.
  • Free of oxidative additives
  • Contains phosphates and non-ionic surfactants
  • Provides high degree of material protection
  • Ideal for mechanical cleaning of stainless steel instruments, utensils and laboratory glassware
Dosage: 1 to 4 g/l
Caution: For avoiding water stains the use of deionised water in the final rinse is recommended.
ORDER
Expand All / Collapse All
Description Packed Pk VWR Catalog Number Quantity
neodisher® MA akaline detergent 10 kg bucket 10 kg MIELUS486876
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty