Potassium sulphate, anhydrous, granular, AR® low in nitrogen, Macron Fine Chemicals™
Management of Change (MOC) category = N
7148-06EA
74063
INR
7148-06
Potassium sulphate, anhydrous, granular, AR® low in nitrogen, Macron Fine Chemicals™
Potassium sulphate
Formula:
K₂SO₄ MW: 174,26 g/mol Boiling Pt: 1698 °C (1013 hPa) Melting Pt: 1069 °C Density: 2,66 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 12,07845 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Chloride (Cl) | ≤ 0.1 % |
Identification A | Passes Test |
Identification B | Passes Test |
Total Nitrogen Compounds (as NO₃) | ≤ 0.001 % |
Solubility (5 in 100) | Passes Test |
Thru U.S. No. 30 Sieve | ≥ 99 % |
On U.S. No. 80 Sieve | ≥ 98 % |
On U.S. No. 100 Sieve | ≥ 99 % |
Appearance (fine, dry, white free-flowing crystalline granules) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...