Calcium hydroxide, powder, BAKER ANALYZED® ACS, J.T.Baker®
1372-05EA
0
INR
1372-05
Calcium hydroxide, powder, BAKER ANALYZED® ACS, J.T.Baker®
Calcium hydroxide
Formula:
Ca(OH)₂ MW: 74,09 g/mol |
MDL Number:
MFCD00010901 CAS Number: 1305-62-0 UN: 3262 ADR: 8,III |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (Ca(OH)₂) | ≥95.0 % |
Calcium Carbonate (CaCO₃) | ≤3.0 % |
Insoluble in HCl | ≤0.03 % |
Chloride (Cl) | ≤0.03 % |
Sulfur Compounds (as SO₄) | ≤0.1 % |
Iron (Fe) | ≤0.05 % |
Heavy Metals (as Pb) | ≤0.003 % |
Magnesium (Mg) | ≤0.5 % |
Potassium (K) | ≤0.05 % |
Sodium (Na) | ≤0.05 % |
Strontium (Sr) | ≤0.05 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...