Barium sulphate 97.5-100.5% USP, Macron Fine Chemicals™
8821-04EA
8440
INR
8821-04
Barium sulphate 97.5-100.5% USP, Macron Fine Chemicals™
Barium sulphate
Formula:
BaSO₄ MW: 233,39 g/mol Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 1345 °C Density: 4,5 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003455 CAS Number: 7727-43-7 EINECS: 231-784-4 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (BaSO₄) | 97.5 - 100.5 % |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - pH of(10% Aqueous Solution) | 3.5 - 10.0 |
USP - Acid-Soluble Substances | ≤0.3 % |
USP - Soluble Barium Salts | ≤0.001 % |
Bulkiness (ml) | ≥11 |
USP - Sulfide (µg/g) | ≤0.5 |
Appearance (fine, white powder) | |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...