Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Acid red 18 ≥75%, solid CI 16255

Supplier: PanReac AppliChem
Synonyms: Cochineal Red APonceau 4RCochenille redFood Red No. 102Scarlet 3RNew Coccine

A4291.0025EA 0 INR InStock
A4291.0025
Acid red 18 ≥75%, solid CI 16255
Acid red 18


Formula: C₂₀H₁₄N₂O₁₀S₃
MW: 604,46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004084
CAS Number: 2611-82-7
Order Now
Specification Test Results
Assay Min. 75%
UV spectrum Passes test
Form Solid

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty