Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staining solutions and kits, Gurr®

Supplier: VWR Chemicals
Gurr®
The Gurr range provides the first truly international standard for microscopical dyes and stains.

Gurr specifications have been devised by combining practical biological testing with absolute chemical criteria.



 

ORDER

Expand All / Collapse All
Description Pack type Pk VWR Catalog Number Quantity
Elastin stain (Miller) (form. R.A. Lamb) Glass bottle 500 ml 351154S
Get a discount if you order 6 or more. Login in now for details.
Eosin yellowish for microscopical staining Gurr® Glass bottle 100 g 341973R
Get a discount if you order 6 or more. Login in now for details.
Eosin yellowish for microscopical staining Gurr® Glass bottle 1 kg 341975T
Get a discount if you order 6 or more. Login in now for details.
Eosin yellowish for microscopical staining Gurr® Glass bottle 25 g 341972Q
Get a discount if you order 6 or more. Login in now for details.
Feulgen stain (Schiff) (form. R.A. Lamb) Glass bottle 500 ml 351204L
Get a discount if you order 6 or more. Login in now for details.
Giemsa`s stain improved R66 solution Gurr® Glass bottle 1 l 350865P
Get a discount if you order 6 or more. Login in now for details.
Giemsa`s stain improved R66 solution Gurr® Glass bottle 500 ml 350864X
Get a discount if you order 6 or more. Login in now for details.
Giemsa`s stain improved R66 solution Gurr® Metal drum 25 l 35086HE
Giemsa`s stain solution Glass bottle 500 ml 352603R
Get a discount if you order 6 or more. Login in now for details.
Haemalum (Mayer`s) Gurr® for microscopical staining Glass bottle 500 ml 350604T
Get a discount if you order 6 or more. Login in now for details.
Haematoxylin (monohydrate) for microscopical staining Glass bottle 25 g 340374T
Get a discount if you order 6 or more. Login in now for details.
Haematoxylin Harris (mercury-free) - 1 l 351945S
Get a discount if you order 6 or more. Login in now for details.
Hema Gurr® rapid staining set for haematology Glass bottle 1 351042L
Leishman`s staining solution Gurr® for microscopical staining Glass bottle 500 ml 350224L
Get a discount if you order 6 or more. Login in now for details.
Lugol`s iodine solution Glass bottle 250 ml 351903Y
Get a discount if you order 6 or more. Login in now for details.
May-Grunwald`s eosin methylene blue solution Glass bottle 1 l 352065W
Get a discount if you order 6 or more. Login in now for details.
May-Grunwald`s stain solution Glass bottle 1 l 352622M
Get a discount if you order 6 or more. Login in now for details.
May-Grunwald's stain (form. RA Lamb) Glass bottle 1 l 351355C
Get a discount if you order 6 or more. Login in now for details.
Methylene blue for microbiological staining Gurr® and reagent for molybdate Glass bottle 25 g 340484B
Get a discount if you order 6 or more. Login in now for details.
Neutral red for microscopical staining Gurr® Glass bottle 100 g 340565B
Neutral red for microscopical staining Gurr® Glass bottle 25 g 340564A
Get a discount if you order 6 or more. Login in now for details.
Nuclear fast red for microscopical staining Gurr® Glass bottle 25 g 342094W
Get a discount if you order 6 or more. Login in now for details.
Papanicolaou`s stain OG 6 Gurr® Plastic bottle 1 l 350405X
Get a discount if you order 6 or more. Login in now for details.
Papanicolaou EA 50 (new formulation) Glass bottle 1 l 351695T
Get a discount if you order 6 or more. Login in now for details.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty