1536-well (UHTS) microplates for cell culture, BRANDplates®
Supplier: Brand
735-2113EA 0 INR
BRND782000 BRND782002 BRND782001
1536-well (UHTS) microplates for cell culture, BRANDplates®
PS. Cell culture treated flat bottomed plates.
cellGrade™: The standard surface for cultivation of adherent cell lines. Chemical groups on the surface of the plate bind to serum compounds, stimulating the growth of immobilised cells.