  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Carrez I solution potassium hexacyanoferrate(II)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR Chemicals
Description:   Carrez I solution potassium hexacyanoferrate(II) 0.25 mol/l in aqueous solution
UOM:  1 * 1 l
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com