Ciprofloxacin hydrochloride 98-102% (by HPLC on dried basis) Ph. Eur., pharma grade
Synonyms:
1-Cyclopropyl- 6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride
, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
A4556.0005EA
0
INR
A4556.0005
Ciprofloxacin hydrochloride 98-102% (by HPLC on dried basis) Ph. Eur., pharma grade
Ciprofloxacin hydrochloride
Certifications: Meets the requirements of the European Pharmacopoeia (Ph. Eur. 8.4).
Formula:
C₁₇H₁₈FN₃O₃·HCl MW: 367,84 g/mol |
MDL Number:
MFCD00079044 CAS Number: 86393-32-0 |
Specification Test Results
State of Matter | Solid |
Assay (HPLC, calc. on dried subst.) | 98,0 - 102,0% |
Appearance of solution | Passes Test |
Identity | Passes Test |
pH (2,5%; H2O) | 3,5 - 4,5 |
Heavy metals (as Pb) | Max. 0,002 % |
Sulfated ash | Max. 0,1% |
Related subst. (HPLC) | Passes Test |
Fluoroquinolinic acid | Max. 0,2% |
Water (K.F.) | Max. 6,7% |
Storage | Store at 2 to 8 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...