ICNA04807426
ICNA04807426EA 3678 INR
ICNA04807426 ICNA04807423
ICNA04807426
Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.
The principal uses of Triton® X-100 is in industrial and household detergent applications and in emulsifying agents. It is employed in almost every type of liquid, paste, and powdered cleaning compound, ranging from heavy-duty industrial products to gentle detergents for fine fabrics. It is also important ingredient of primary emulsifier mixtures used in the manufacture of emulsion polymers, stabilizers in latex polymers, and emulsifiers for agricultural emulsion concentrates, and wettable powders.
Cloud point=63 - 69 °C(1% aq soln); Pour Point °F=45; HLB Value=13.5; Average EO Units=9-10
Miscible in Water, Polar Organic Solvents (alcohols, glycols, ethers, ketones, etc.) & Aromatic Hydrocarbons; Soluble in Inorganic Salt Solutions & Aqueous Mineral Acids; Insoluble in Aliphatic Hydrocarbons.
Caution: Research Use Only (RUO).