Tube rack, customisable
525-0949EA 3424 INR
VWRI525-0949
Tube rack, customisable
This multi-purpose test tube rack is designed with the flexibility to hold 20×5 ml and 20×15 ml tubes, or to hold 40×5 ml or 40×15 ml tubes.
  • Teardrop shaped openings (top tray) for better retention of tubes
  • Ergonomic handles for ease of use
  • Imprinted grid on both sides of the upper shelves for quick identification and retrieval
  • Shelves can be easily rotated to accommodate different size tubes
  • Autoclavable at 121 °C for 15 min
The rack incorporates two moving hinged upper shelves that can be rotated to an up or down position to accommodate the size of the tube being used. In addition, the rack can also hold clinical tubes of a similar diameter (16 mm) and height.