Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Boron Standards

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   B(OH)₃ in H₂O tr.NH₄OH
Supplier:  VWR Chemicals
Description:   Boron standard solution, 1,000 mg/l B in water AVS TITRINORM® standard for AAS
Supplier:  VWR Chemicals
Description:   Boron 1000 ppm ARISTAR mono-element Standard for ICP-MS 1 * 100 ml
Supplier:  Avantor Performance Materials
Description:   Boron standard solution for AAS are prepared from metals and salts of 99,999% spectral purity.
UOM:  1 * 150 ml
Supplier:  VWR Chemicals
Description:   This certified reference material is produced using a high-purity starting material, acid from sub-boiling and 18 MOhm deionised water.
Supplier:  VWR Chemicals
Description:   H₃BO₃ in H₂O tr. NH₄OH
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 6  of 6
 

a new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping