Bluetooth® temperature and humidity loggers
VWRI620-2464 VWRI620-2463
Bluetooth® temperature and humidity loggers
These data loggers are available with or without hygrometer, recording temperature or temperature and humidity data for 30 days. Hourly data recorded for current 7 days, then maximum and minimum temperatures or temperatures and humidity are recorded for next 23 days.
  • Monitor critical environments without a physical connection; maximum signal distance is approx. 40 metres
  • Alarm sounds if temperature falls outside of the set parameters
  • Data viewable as a chart or graph, or on a calendar; data from up to six data loggers can be plotted on one graph for direct comparison
  • Indicators include WBGT (Wet Bulb Global Temperature), Heat, Mold and Comfort indices as well as memory full, active alarm state and low battery
  • Features all-weather housing with internal temperature sensor and humidity sensor
Certifications: Conforms to ROHS compliance and CE marked.
Delivery information: Supplied with Certificate of Compliance indicating accuracy for four temperature points or four temperature and three humidity points, two AAA batteries, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions.