The Yeast Genomic DNA Purification Kit yields high molecular weight DNA from yeast using a non-enzymatic lysis buffer and no glass beads. The protocol is simple, requiring about 1 hour to complete, and may be scaled for large or small samples. The purification process begins with lysis of the yeast cell wall with optional RNase A treatment, and is followed by protein precipitation and then DNA precipitation. Unlike many conventional yeast genomic DNA purification methods, no phenol, chloroform or columns are required.
- Rapid purification yields high quality, high molecular weight DNA
- No enzymes, glass beads or freeze thawing for cell lysis
- Method does not require phenol, chloroform or columns
- Simple, 1 hour protocol which can be easily scaled for large or small samples
The purified DNA yields vary for different yeast strains and is also dependent on cell density. The kit is optimised for Saccharomyces cerevisiae, with an average yield of 1,5 µg DNA per 5×107 cells as measured by the Fluorescent DNA Quantitation Kit. High quality DNA obtained with the Yeast Genomic DNA Purification Kit is compatible downstream applications, such as, PCR, restriction enzyme digestions and Southern blotting.
Delivery information: Kit includes Yeast Lysis Solution, Ribonuclease A (RNase A), Protein Precipitation Solution, TE, pH 8,0.