Microtiter tubes
Supplier: Thermo Fisher Scientific
MLBP845EA 0 INR
MLBPRC-845-TPS MLBP845
Microtiter tubes
Tubes Microtubes
PP, superior grade, microtiter tubes for harvesting cells, cell uptakes and receptor binding.
  • Can be autoclaved, sealed and frozen for storage
  • Can be frozen to -80 ˚C
Ideal for a wide variety of laboratory procedures, such as HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control, blood bank sample freezer storage, transport of specimens and reagents.