Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Sulphanilic acid in acetic acid USP Test Solution (TS)

Supplier: VWR Chemicals
Warning
Synonyms: 4-Aminobenzenesulphonic acidAniline-4-sulphonic acid

85375.180EA 13714 INR InStock
85375.180 85375.260
Sulphanilic acid in acetic acid USP Test Solution (TS)
Sulphanilic acid
Ready to use test solutions (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available


Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 279 °C
Density: 1,485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 0
UN: 2790
ADR: 8,III
Merck Index: 12,09096
https://in.vwr.com/stibo/web/std.lang.all/76/05/2297605.jpg
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty