Calcium sulphate saturated solution in water USP Test Solution (TS)
85339.180EA
1886
INR
85339.180
85339.260
Calcium sulphate saturated solution in water USP Test Solution (TS)
Calcium sulphate
Ready to use test solution (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
Formula:
CaO₄S MW: 136,14 g/mol Melting Pt: 1350 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 Merck Index: 13,01711 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...