You Searched For: Transfer Buffers


3  results were found

SearchResultCount:"3"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Chemicals
Description: Rapid transfer buffer is a simple, single component system for quick and efficient transfer of proteins from SDS-PAGE gels to membranes for Western bl...
Catalog Number: (M115-40L)
Supplier: VWR Chemicals
Description: A pre-blended Tris-glycine powder for preparation of Laemmli SDS-PAGE running buffer. The 40 litre pack size prepares 40 litres of 1X Tris-glycine sol...
UOM: 1 * 40 L

MSDS


Catalog Number: (K872-500ML)
Supplier: VWR Chemicals
Description: For transfer of protein to membranes for sequencing or blotting.
UOM: 1 * 500 mL

MSDS


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 3 of 3
no targeter for Bottom