Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Stacking Buffers

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"2"
  List View Easy View
Sort by:
 
 
 
 


Supplier:  VWR Chemicals
Description:   Bis-Tris Gel Buffer (4X) is used to prepare polyacrylamide gels for optimal separation of small- to medium-sized proteins under denaturing conditions....
UOM:  1 * 1 items
Catalog Number: (M198-500ML)

Supplier:  VWR Chemicals
Description:   This 4X concentrated buffer (0,5 M Tris pH 6,8 and 0,4% SDS) is for the preparation of stacking gels for Laemmli SDS-Page. Dilution to a 1X solution r...
UOM:  1 * 500 ml
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 2  of 2
 

a new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping