Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-4-chlorobenzothiazole hydrobromide ≥97%

Supplier: Alfa Aesar

Purity: 97%
CAS No.: 27058-83-9
Molecular Formula: C7H5ClN2S.HBr
Formula Weight: 265.57
Melting Point: 240-242°
MDL No.: MFCD00035186


Formula: C₇H₅ClN₂S·HBr
MDL Number: MFCD00035186
CAS Number: 27058-83-9


 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
25 g L05440.14
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 240-242°C
Formula weight 265.57
CAS No. 27058-83-9
Purity 97%
Molecular formula C7H5ClN2S.HBr
MDL No. MFCD00035186
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty