(R)-(+)-1-Boc-2-cyanopyrrolidine 97%

Warning

Synonyms: (R)-(+)-1-Boc-2-pyrrolidinecarbonitrile

H57745.03EA 0 INR
H57745.03
(R)-(+)-1-Boc-2-cyanopyrrolidine 97%
(R)-(+)-1-Boc-2-cyanopyrrolidine
Formula: C₁₀H₁₆N₂O₂
MW: 196,25 g/mol
Melting Pt: 32…37 °C
Storage Temperature: Ambient
MDL Number: MFCD06411382
CAS Number: 228244-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR