Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(3-tert-Butylbenzyl)-4-methoxybenzenesulfonamide ≥97%

Supplier: Alfa Aesar

Purity: 97%
Molecular Formula: C18H23NO3S
Formula Weight: 333.44
MDL No.: MFCD22682917



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g H57686.03
250 mg H57686.MD
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula Weight 333.44
Purity 97%
Molecular Formula C18H23NO3S
MDL No. MFCD22682917
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty