Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxetan-3-yl 4-methylbenzenesulfonate ≥96%

Supplier: Alfa Aesar

Purity: 96%
CAS No.: 26272-83-3
Molecular Formula: C10H12O4S
Formula Weight: 228.27
Flash Point: 179°(354°F)
Boiling Point: 372°
MDL No.: MFCD08544401


Formula: C₁₀H₁₂O₄S
MW: 228,27 g/mol
MDL Number: MFCD08544401
CAS Number: 26272-83-3


 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
1 g H57569.03
5 g H57569.06
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula weight 228.27
CAS No. 26272-83-3
Purity 96%
Flash point 179°C(354°F)
Boiling point 372°
Molecular formula C10H12O4S
MDL No. MFCD08544401
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty