tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%
|
Warning
|
Synonyms:
1-BOC-3-Pyrrolidinemethanol
, tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate
H57525.03EA
0
INR
H57525.03
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥97%
tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate
Formula:
C₁₀H₁₉NO₃ MW: 201,27 g/mol Boiling Pt: 285…291 °C Density: 1,089 Storage Temperature: Ambient |
MDL Number:
MFCD02179040 CAS Number: 114214-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...