3-(3-Nitrophenyl)-1H-pyrazole 97%

Warning

H32498.MEEA 0 INR
H32498.ME
3-(3-Nitrophenyl)-1H-pyrazole 97%
3-(3-Nitrophenyl)-1H-pyrazole
Formula: C₉H₇N₃O₂
MW: 189,18 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00106146
CAS Number: 59843-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR