Potassium phenoxymethyltrifluoroborate 95%
|
Warning
|
Synonyms:
Potassium trifluoro(phenoxymethyl)borate
H32030.03EA
0
INR
H32030.03
Potassium phenoxymethyltrifluoroborate 95%
Potassium phenoxymethyltrifluoroborate
Formula:
C₇H₇BF₃KO MW: 214,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10566516 CAS Number: 1027642-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...