Would you like to log in again?
Test kits, triazole
Supplier: Hach Lange
TZ-1 Kit provides proper maintenance and economy of operation of modern boilers and cooling towers which requires balance between water quality parameters and treatment chemicals. It measures common corrosion and biological growth inhibitor using simple color disc method.
Delivery information: Includes benzotriazole and tolyltriazole color discs, comparator box and tubes, reaction bottle, reagents, UV lamp, 230 V power supply, UV safety goggles, instructions and carrying case.
ORDER
Expand All / Collapse All
Description Range Pk VWR Catalog Number Quantity
Triazole test kit TZ-1 0 -15 mg/l 1 KIT HACH2167502
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty