Would you like to log in again?
Bottles, wide neck, with thread
Supplier: RIXIUS
Glass, amber
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes. Amber coloured to protect light-sensitive materials.
Ordering information: Screw caps must be ordered seperately.
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Capacity Thread Ø ext. Height Pk VWR Catalog Number Quantity
100 ml GL 45 48,50 mm 85,50 mm 63 RIXI1-0301-0100-45
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Expand All / Collapse All
Description Thread Pk VWR Catalog Number Quantity
Screw cap, PP, black, with LDPE inlay GL 45 100 215-1926
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty