Would you like to log in again?
Test kits, professional boiler treatment/boiler feed and cooling water
Supplier: Hach
These kits are used for controlling boiler and cooling water treatement. This kit tests for alkalinity, chloride, hardness, sulfite, phopsphate, pH, iron, silica, chlorine, nitrite and total dissolved solids (TDS).
  • Simple, economical methods
  • All-inclusive, the reagents are supplied with the kit
Delivery information: Includes thirty-two reagents, two flasks, two cylinders, one test tube clamp, clippers, four stoppers, filter paper, three droppers, two delivery tubes, one digital titrator, one heatab cookit folding stove with fuel, a demineralizer bottle, a TDS tester, color comparator box, viewing adapter, eight color discs, four mixing bottles, a manual, and carrying case.
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact VWR for details.
ORDER
Expand All / Collapse All
Description Range Pk VWR Catalog Number Quantity
Professional boiler treatment/boiler feed and cooling water test kit, Model PBC-DT Phosphate 0 - 40 mg/l; pH 7,4 - 9,6; Fe 0 - 2,0 mg/l; Silica 0 - 30 mg/l; Chlorine 0 - 3,4 mg/l; Nitrite 0 - 80 mg/l 1 KIT HACH2350700
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty