Thymol blue, BAKER ANALYZED® ACS, J.T. Baker®

Supplier: Avantor
Warning

Synonyms: Thymolsulphonphthalein

V856-01EA 7715.77 INR
V856-01 V856-02
Thymol blue, BAKER ANALYZED® ACS, J.T. Baker®
Thymol blue

Lot analysis on label.

Formula: C₂₇H₃₀O₅S
MW: 466,6 g/mol
Melting Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Clarity of Solution Passes Test
Visual Transition Intervals - pHÖ9.2 (Blue) Passes Test
Visual Transition Intervals - pHÖ1.2 (Pink) Passes Test
Visual Transition Intervals - pHÖ2.8 (Yellow) Passes Test
Visual Transition Intervals - pHÖ8.0 (Yellow) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR