Tetrabutylammonium bromide 90-100%, J.T.Baker®
V356-05EA
16825
INR
V356-05
Tetrabutylammonium bromide 90-100%, J.T.Baker®
Tetrabutylammonium bromide
Formula:
C₁₆H₃₆BrN MW: 322,37 g/mol Melting Pt: 100…103 °C Density: 1,148 g/cm³ (20 °C) |
MDL Number:
MFCD00011633 CAS Number: 1643-19-2 EINECS: 216-699-2 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
4% Stacking Gel Components Mix | |
6% Running Gel* | |
(pH 8.8, approx. 0.375M) | |
*The above values are the results of the proper mixing of the 2 kit components. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...