Ninhydrin, BAKER ANALYZED® ACS, J.T.Baker®
|
Warning
|
Synonyms:
2,2-Dihydroxyindan-1,3-dion
, Trioxohydrindene monohydrate
, Ninhydrin monohydrate
, 2,2-Dihydroxy-1,3-indanedione
, Indanetrione hydrate
, 2,2-Dihydroxyindane-1,3-dione
, 1,2,3-Indantrione monohydrate
N862-03EA
20386.01
INR
N862-03
Ninhydrin, BAKER ANALYZED® ACS, J.T.Baker®
Ninhydrin
Lot analysis on label.
Formula:
C₉H₆O₄ MW: 178,14 g/mol Boiling Pt: 351 °C (1013 hPa) Melting Pt: 250…258 °C Density: 0,862 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003791 CAS Number: 485-47-2 EINECS: 207-618-1 Merck Index: 12,06646 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Identification and Melting Point | Passes Test |
Solubility | Passes Test |
ACS - Sensitivity to Amino Acids | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...