Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrocatechol, J.T. Baker®

Supplier: Avantor Performance Materials
Warning

U672-07EA 16790 INR InStock
U672-07
Pyrocatechol, J.T. Baker®
Pyrocatechol


Formula: C₆H₆O₂
MW: 110,11 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 103…105 °C
Density: 1,0548 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1,III
Merck Index: 12,08183
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Melting Point 102 - 106 °C

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty