1-Octanesulphonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Warning
|
Synonyms:
Sodium octane-1-sulphonate
H165-56EA
0
INR
H165-56
1-Octanesulphonic acid sodium salt, monohydrate ≥98%, OR, Macron Fine Chemicals™
1-Octanesulphonic acid sodium salt
Powder. For use in PIC and HPLC applications.
Formula:
C₈H₁₇NaO₃S MW: 216,28 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007544 CAS Number: 5324-84-5 EINECS: 226-195-4 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (CH₃(CH₂)₇SO₃Na·H₂O) | ≥98.0 % |
Solubility | Passes Test |
Optical Density at 210 nm (au) | ≤0.05 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...