Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.

Thiourea ≥99% ACS, J.T. Baker®

Supplier: Avantor Performance Materials
Warning
Synonyms: ThiocarbamideSulfourea

4123-05EA 0 INR InStock
4123-05
Thiourea ≥99% ACS, J.T. Baker®
Thiourea
Lot analysis on label.


Formula: NH₂CSNH₂
MW: 76,12 g/mol
Melting Pt: 174…177 °C
Density: 1,435 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
EINECS: 200-543-5
UN: 3077
ADR: 9,III
Merck Index: 12,09505
https://in.vwr.com/stibo/web/std.lang.all/72/67/2297267.jpg
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty