Cadmium acetate dihydrate, crystals, BAKER ANALYZED®, J.T.Baker®
|
Warning
|
1190-04EA
14225
INR
1190-04
Cadmium acetate dihydrate, crystals, BAKER ANALYZED®, J.T.Baker®
Cadmium acetate dihydrate
Formula:
(H₃CCOO)₂Cd·2H₂O MW: 266,53 g/mol Boiling Pt: 105…107 °C (4 mmHg) Melting Pt: 256 °C Density: 2,341 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150020 CAS Number: 5743-04-4 EINECS: 208-853-2 UN: 2570 ADR: 6.1,III Merck Index: 13,01614 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Insoluble Matter | ≤ 0.005 % |
Chloride (Cl) | ≤ 0.001 % |
Nitrate (NO₃) | ≤ 0.003 % |
Sulfate (SO₄) | ≤ 0.005 % |
Copper (Cu) | ≤ 0.001 % |
Trace Impurities - Iron (Fe) | ≤ 0.001 % |
Lead (Pb) | ≤ 0.003 % |
Zinc (Zn) | ≤ 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...