Formamide ≥98%, OR, Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
Synonyms:
Carbamaldehyde
3797-08EA
0
INR
3797-08
Formamide ≥98%, OR, Macron Fine Chemicals™
Formamide
Formula:
HCONH₂ MW: 45,04 g/mol Boiling Pt: 210 °C (1013 hPa) Melting Pt: 2 °C Density: 1,1334 g/cm³ (20 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 Merck Index: 13,04261 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (HCONH₂) | ≥98 % |
Color (APHA), For Information Only | |
Specific Gravity at 25°/25°C, For Information Only | |
Appearance (clear, colorless to pale yellow solution) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...