Desiccators, vacuum, Desi-Vac™
VWRU62344-930
Desiccators, vacuum, Desi-Vac™
These desiccators are ideal for storing chemicals, powders, sample tissues, plant sections, weighing, drying, preserving, storing and even shipping samples.
  • Rubber O-ring-sealed cover maintains vacuum
  • Achieve a vacuum of better than 200" of water with built-in pump
  • Vacuum pump built into the top removes air with manual strokes
  • Release key on top allows air into the container to remove lid