VWR®Signature™, Incubators, reach-in
Supplier: VWR
VWR®
VWRU35962-096EA 748922 INR
VWRU35962-062 VWRU35962-096
VWR®Signature™, Incubators, reach-in
Incubators Standard Incubators
These incubators provide gentle air flow and minimises the space. And the chamber reinforced to support roller apparatus and shakers.
  • Microprocessor temperature control
  • Over temperature safety protection
  • Temperature uniformity to 0,8 °C
  • Prewired access panel for chart recorder
The air jacket design and heated outer door allows for uniform heat throughout the incubators. Both models incorporate a microprocessor controller to achieve precise temperature uniformity. An independent, secondary temperature controller offers the added security of over temperature protection. The chamber floors are reinforced to support roller apparatus or shakers.
Delivery information: Supplied with six shelves.