Pinch cock, heavy duty, Talon®
VWRU21573-650EA 1941 INR
VWRU21573-650
Pinch cock, heavy duty, Talon®
This pinch cock with heavy duty spring is the perfect choice when using stiff tubing common in gas and water connections or when you need more pressure.
  • Ideal for start/stop precision
  • Offers simple, one-handed operation and quick, convenient, complete closure when placed on tubing and released
  • Won't cut tubing
  • Corrosion- and rust-resistant
  • For heavy-wall tubing up to 11 mm (⁷/₁₆") Ext.Ø.