Pinch cock, regular, Talon®
VWRU21573-155EA 1990 INR
VWRU21573-155
Pinch cock, regular, Talon®
This regular pinch cock offers simple, one-handed operation and quick, convenient, complete closure when placed on tubing and released.
  • Won't cut tubing
  • Corrosion- and rust-resistant
  • For tubing up to 12 mm (¹/₂") Ext.Ø.