These manometers with ABS casing display differential pressure or vacuum readings in 11 different units: psi, mbar, bar, cm/H₂O, in./H₂O, ft./H₂O, mm/Hg, in./Hg, kg/cm², oz./in.², and kPa, with a response time of 0,5 seconds.
- Simple hose fittings permit use of tubing with inside diameter from 0,63 to 1,9 mm (¹/₁₆ to ³/₁₆")
- Large 6,3 mm (⁵/₈") high digits are visible from 1,8 m (6') away; brilliant backlighting permits reading even in dark areas
- Unit may be zeroed with the touch of a button
- Differential feature registers the pressure or vacuum difference between two different sources
- Memory feature recalls highest and lowest readings over any time period
- Count-up timer displays times up to 99 hours, 59 minutes, 59 seconds
- Data hold key freezes the display to capture readings
Compliance with ISO/IEC 17025:2005 (1750.01); accredited by A2LA indicating traceability to standards provided by NIST.
Delivery information: Supplied with hose/tubing, 9 V alkaline battery, and RS-232 serial computer output.