Aerosol duster, Whoosh-Duster™
VWRU16650-027
Aerosol duster, Whoosh-Duster™
Whoosh-Duster™ is a portable, noncorrosive, nonflammable, dry and odorless gas. Contains eleven ounces of pure compressed gas, triple-filtered to 0,3 microns to eliminate moisture and all contaminants. The "air compressor in a can" provides up to 3000 jets of cleaning power. Ideal for clean optical lenses, balance parts, microscope slides, relay contacts, spectrophotometer cells, photographic slides, circuit boards, syringes, micropipettes and flame atomisers.
  • Instant blows of lint, dust and dirt with 4,9 bar of high pressure, leaving zero residue
  • Trigger actuator valve provides a controlled delivery
  • Duster contains an EPA-designated substitute for ozone-depleting HFC's and CFC's