VWR®, Stoppers, Gum Rubber, with 1 Hole
Supplier: VWR
VWR®
VWRU59584-354EA 0 INR
VWRU59584-274 VWRU59584-354
VWR®, Stoppers, Gum Rubber, with 1 Hole
Stoppers Rubber Stoppers
These stoppers contain less free sulphur and have a lower density than black rubber stoppers. Its composition ensures high resilency and holes facilitates the laboratory usage. Ideal for food grade applications such as beer and wine making used with carboys.
  • Natural gum rubber
  • White