VWR®, Beaker Tongs, Carbon Steel
Supplier: VWR
VWRU82027-360EA 0 INR
VWRU82027-360
VWR®, Beaker Tongs, Carbon Steel
Tongs
Carbon steel, cadmium plated, with plastic coated tips.
Designed for the safe and simple handling of beakers and similar vessels from 50 to 2000 ml capacities.