Cell culture bottles, Lexan®, VWR®
VWRU89107-028EA 0 INR
VWRU89107-028
Cell culture bottles, Lexan®, VWR®
PC, wide mouth.
  • Withstand maximum speeds without leakage
  • Tightfitting polypropylene screw caps reduces aerosol escape
  • Optically clear and shatterproof
  • Autoclavable
These bottles are ideal for applications that include culture of microbes, plants, algae, and organisms requiring large surface area to volume ratios and also in ultracentrifugation process. Mostly used where relatively small amounts of solids are involved.