Metallographic camera, VisiCam® MET 5000
630-2669EA 280523 INR
VWRI630-2669
Metallographic camera, VisiCam® MET 5000
Scientific camera that delivers a high frame rate (30 fps at full resolution) and stable image transmission.
  • Compact and dust-proof
  • Low noise and excellent colour reproduction
  • Observation methods: Brightfield and Darkfield
Ideal for applications such as semiconductor inspection, material science, defect analysis, microscopy, medical engineering, quality assurance and live cell imaging.
Standard C-mount attachment allows the camera to be attached to trinocular microscopes (provided the right C-mount adapter is used) or to binocular/monocular microscopes (with the optional eyepiece adapter 630-1121).
Ordering information: Includes USB cable and professional image analysis software. Additional software functions available for grain size analysis, cast iron analysis, non metallic inclusion rating, rust grade and phase analysis. Please contact your local VWR sales office for further information.