Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Supplier: VWR
Avantor® ACE®
ACE-1210-35005EA 42019 INR
ACE-1210-35005 ACE-1210-0101GD ACE-1210-05005 ACE-1210-12005 ACE-1210-30005 ACE-1210-03005S ACE-1210-0501 ACE-1210-10005 ACE-1210-03005 ACE-1210-12005S ACE-1210-25005 ACE-1210-25005S ACE-1210-10005S ACE-1210-75005S ACE-1210-15005S ACE-1210-15005 ACE-1210-05005S ACE-1210-75005
Avantor® ACE® C18-PFP, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C18-PFP phase utilises a specially developed ligand combining a C18 chain with integral PFP functionality. This results in a phase that maintains the hydrophobic, stability and low bleed characteristics of leading C18 phases, yet provides the multiple retention mechanisms of a PFP phase.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 µm to 10 µm) and complementary phases
  • Also useful for analytes with π-bonding or electron or proton donor groups
  • Ultra-low bleed to ensure LC-MS compatibility
  • Method validation kits contain three different batches of the same phase - available on request
Due to its integral pentafluorophenyl functionality, Avantor® ACE® C18-PFP is also recommended for separations that involve halogenated aromatic compounds, regioisomers, and those analytes with differing shape constraints.
These columns are made from stainless steel.
Ordering information: Method development and validation kits are available on request. Please contact chromsupport@avantorsciencs.com.