Vacuum aspiration systems, Quicktrap
Supplier: CBS Scientific
CBSCQT-100-5EA 0 INR
CBSCQT-100-5
Vacuum aspiration systems, Quicktrap
Quicktrap vacuum protection system is designed for the aspiration of standard, radioactive or pathogenic supernatants.
  • With acrylic cabinet, primary and secondary traps, tygon tubing, heavy duty Nalgene bottles
Delivery information: It includes tubing, connectors, back-flow filter and bottle liquid storage containers.