Conductivity meters, handheld, CO 300 / LCO 302
Supplier: VWR Collection
663-0222EA 0 INR
VWRI663-0222 VWRI663-0223
Conductivity meters, handheld, CO 300 / LCO 302
These two portable meters are designed for different applications: CO 300 is capable of measuring conductivity, salinity, TDS and temperature values and can be universally used. Typical applications include: Fresh and sea water aquaristics, fish farming, water and drinking water monitoring. LCO 302 measures conductivity, resistivity and temperature and is specifically designed for ultra-pure water applications. It can also be used for functional checks of ion exchangers.
  • Made of impact resistant ABS, front side is IP 65 rated
  • Min./Max. value memory, 'HOLD' function, auto-off function
  • Automatic temperature compensation via integrated temperature sensor
  • Supplied ready-to-use with measuring cells
Delivery information: CO 300 is supplied with 2-pole measuring cell (Ø 12 mm, graphite) with 1,2 m cable, battery, calibration protocol and manual. LCO 302 is supplied with 2-pole measuring cell (Ø 12 mm, stainless steel) with 1,2 m cable, battery and manual.